Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Clear Boston Round Bottles with Black Phenolic Pulp/Vinyl Cap
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
960 mL
Closure Size:
20-400 mm
22-400
24-400 mm
28-400 mm
33-400 mm
Packaging:
Ampule
Bulk pack
Convention pack
Plain carton
Qorpak carton
Quantity:
12/Cs.
24/Cs.
24/Cs., 1 mL
432/Cs.
48/Cs.
60/Cs.
Description
- Features a round body, curved shoulders and narrow screw neck opening
- Environmental sensitive help eliminate waste and ensure product integrity for long term storage
- Glass offers minimal chemical interference with contents
Specifications
Specifications
Capacity (Metric) | 120 mL |
CAS | 64-17-5, 60-33-3 |
Capacity (English) | 4 oz. |
Boiling Point | 229 to 230°C/16 mmHg (lit.) |
Melting Point | -5°C (lit.) |
Color | Clear |
Material | Glass (Type 3) |
Linear Formula | CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H |
MDL Number | MFCD00064241 |
UN Number | UN 1170 - class 3 - PG 2 - Ethanol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction