Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Clear French Square Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Lined Cap
Click to view available options
Capacity (Metric):
15 mL
60 mL
120 mL
240 mL
480 mL
950 mL
Closure Size:
20-400 mm
28-400 mm
33-400 mm
43-400 mm
48-400 mm
58-400
Quantity:
24/Cs.
24/Cs., 1 mL
48/Cs.
Description
Vacuum and Ionization Cleaning Process: Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Next, the air passes through an absorbent filter to remove gases. A vacuum draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the process.
- Features wide mouth
- Clear glass provides maximum content visibility and minimal chemical inference with bottle contents
- Designed to save shelf and storage space
- Attached caps reduce carton dust
Specifications
Specifications
Capacity (Metric) | 950 mL |
CAS | 75-05-8, 197771-77-0 |
Capacity (English) | 32 oz. |
Color | Clear |
Diameter (Metric) Outer Neck | 58 mm |
Material | Glass (Type 3) |
Linear Formula | C17H21NO5 · HCl |
UN Number | UN 1648 3 / PGII |
Concentration | 1 mg/mL in acetonitrile (as free base) |
No. per Case | 24/Cs. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction