Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Clear Serum Bottle, Crimp Top
Click to view available options
Volume (Metric):
2.5 mL
30 mL
50 mL
100 mL
No. per Pack:
144/Pk.
272/Pk.
288/Pk.
36/Pk.
Specifications
Specifications
Description | No Closure Included |
Diameter (Metric) Opening | 20 mm |
Color | Clear |
Diameter (Metric) Outer | 36.6 mm |
Material | Glass |
Volume (Metric) | 30 mL |
No. per Pack | 36/Pk. |
Autoclavable | Autoclavable |
Closure Type | Crimp Cap |
Height (Metric) | 62.8 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction