Learn More
cleavage stimulation factor Antibody (10E11), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | cleavage stimulation factor |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1000000000000 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CF-1 50 kDa subunit, Cleavage stimulation factor 50 kDa subunit, cleavage stimulation factor subunit 1, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa, CSTF 50 kDa subunit, CstF-50, CstFp50 |
| Gene Symbols | CSTF1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.