Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cleavage stimulation factor Antibody (10E11), FITC, Novus Biologicals™
SDP

Catalog No. NBP242659F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242659F 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242659F Supplier Novus Biologicals Supplier No. NBP242659F

Mouse Monoclonal Antibody

cleavage stimulation factor Monoclonal specifically detects cleavage stimulation factor in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen cleavage stimulation factor
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1000000000000
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias CF-1 50 kDa subunit, Cleavage stimulation factor 50 kDa subunit, cleavage stimulation factor subunit 1, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa, CSTF 50 kDa subunit, CstF-50, CstFp50
Gene Symbols CSTF1
Host Species Mouse
Immunogen Recombinant human CSTF1 (1-431aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1477
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.