Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC-1 Antibody (MM0187-8K42), PerCP, Novus Biologicals™
SDP

Catalog No. NB361286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361286 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361286 Supplier Novus Biologicals Supplier No. NBP212192PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC-1 Monoclonal antibody specifically detects CLEC-1 in Human samples. It is validated for Western Blot

Specifications

Antigen CLEC-1
Applications Western Blot
Classification Monoclonal
Clone MM0187-8K42
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CLEC-1, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328
Host Species Mouse
Immunogen Human recombinant CLEC-1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51267
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.