Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC-2/CLEC-1B Antibody (AYP1), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N25016PECY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N25016PECY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N25016PECY7 Supplier Novus Biologicals Supplier No. NBP250166PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC-2/CLEC-1B Monoclonal specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CLEC-2/CLEC-1B
Applications Flow Cytometry
Classification Monoclonal
Clone AYP1
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721
Gene Symbols CLEC1B
Host Species Mouse
Immunogen A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51266
Test Specificity AYP1 can block the interaction of human CLEC-2 with its endogenous ligand Podoplanin. AYP1 does not cross react with mouse CLEC-2 when tested via FACS. AYP1 does not detect CLEC-2 by western blot. For WB we recommend clone AYP2.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.