Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC-2/CLEC-1B Antibody (AYP2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB203238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203238 Supplier Novus Biologicals Supplier No. NBP250167JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC-2/CLEC-1B Monoclonal antibody specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Western Blot

Specifications

Antigen CLEC-2/CLEC-1B
Applications Western Blot
Classification Monoclonal
Clone AYP2
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721
Host Species Mouse
Immunogen A recombinant extracellular fragment of human CLEC-2 (aa 68-229), which is the extracellular domain of the protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51266
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.