Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC-2A Antibody (705518) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB374967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374967 Supplier Novus Biologicals Supplier No. FAB7219J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC-2A Monoclonal antibody specifically detects CLEC-2A in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CLEC-2A
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 705518
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLEC2A C-type lectin domain family 2, member A, KACL, PILAR, UNQ5792
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC-2A, Trp49-Leu174 (predicted), Accession # Q6UVW9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387836
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.