Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC10A/CD301 Antibody (019), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB316248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316248 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316248 Supplier Novus Biologicals Supplier No. NBP290688JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Mouse samples. It is validated for ELISA

Specifications

Antigen CLEC10A/CD301
Applications ELISA
Classification Monoclonal
Clone 019
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CLEC10A/CD301 (Accession#: NP_001191181.1; Gln58-Ser305).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10462
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.