Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC10A/CD301 Antibody (ER-MP23), HRP, Novus Biologicals™
SDP

Catalog No. NB208636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB208636 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB208636 Supplier Novus Biologicals Supplier No. NB10064874H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen CLEC10A/CD301
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-MP23
Conjugate HRP
Formulation PBS
Gene Alias CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL
Host Species Rat
Immunogen Balb/c macrophage precursor cell hybrids
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10462
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.