Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC14A Antibody (036), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB332896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332896 Supplier Novus Biologicals Supplier No. NBP290521APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC14A Monoclonal antibody specifically detects CLEC14A in Mouse samples. It is validated for ELISA

Specifications

Antigen CLEC14A
Applications ELISA
Classification Monoclonal
Clone 036
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGFR5, EGFR-5, Epidermal growth factor receptor 5
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse CLEC14A (Accession#: NP_080085.3, Met1-Thr386)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161198
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.