Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC18C Antibody (118F2.02), Novus Biologicals™
SDP

Catalog No. DDX0381P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0381P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0381P100 Supplier Novus Biologicals Supplier No. DDX0381P100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC18C Monoclonal specifically detects CLEC18C in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CLEC18C
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 118F2.02
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias C-type lectin domain family 18 member C, C-type lectin domain family 18, member C, mannose receptor-like 3, Mannose receptor-like protein 3, MGC34761, MRCL, MRCL3, MRLP3
Gene Symbols CLEC18C
Host Species Mouse
Immunogen Mannose receptor-like/CD206-like was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283971
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.