Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC2L Antibody (2326G), PerCP, Novus Biologicals™
SDP

Catalog No. NB305859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305859 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305859 Supplier Novus Biologicals Supplier No. FAB10204C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC2L Monoclonal specifically detects CLEC2L in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CLEC2L
Applications Flow Cytometry
Classification Monoclonal
Clone 2326G
Conjugate PerCP
Formulation PBS
Gene Alias C-type lectin domain family 2 member L, C-type lectin domain family 2, member L
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human CLEC2L protein, Ser90-Thr214, Accession # P0C7M8-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154790
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.