Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC3B/Tetranectin Antibody (006), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB316791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316791 Supplier Novus Biologicals Supplier No. NBP290147APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC3B/Tetranectin Monoclonal antibody specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CLEC3B/Tetranectin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 006
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CLEC3B/Tetranectin (Accession#: AAX37102.1; Met1-Val202).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7123
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.