Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC3B/Tetranectin Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB104043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104043 Supplier Novus Biologicals Supplier No. NBP298345AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLEC3B/Tetranectin Polyclonal specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CLEC3B/Tetranectin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CLEC3B/Tetranectin (P43025; Met1-Val202).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7123
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.