Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC3B/Tetranectin Rabbit anti-Mouse, Clone: 11, Novus Biologicals™
SDP

Catalog No. NB025116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025116 50 μg
NB025115 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025116 Supplier Novus Biologicals Supplier No. NBP29068450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC3B/Tetranectin Monoclonal specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for ELISA.

Specifications

Antigen CLEC3B/Tetranectin
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
Host Species Rabbit
Immunogen Recombinant Mouse CLEC3B/Tetranectin protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7123
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.