Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4D/CLECSF8 Antibody (413512), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB364120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364120 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364120 Supplier Novus Biologicals Supplier No. FAB2806E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4D/CLECSF8 Monoclonal antibody specifically detects CLEC4D/CLECSF8 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CLEC4D/CLECSF8
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 413512
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8, Gly52-Asn215, Accession # Q8WXI8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338339
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.