Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4D/CLECSF8 Antibody (WEN42), Novus Biologicals™
SDP

Catalog No. NB119818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119818 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119818 Supplier Novus Biologicals Supplier No. NBP311193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4D/CLECSF8 Monoclonal specifically detects CLEC4D/CLECSF8 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional.

Specifications

Antigen CLEC4D/CLECSF8
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone WEN42
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL
Host Species Mouse
Immunogen BWZ.36 cell expressing chimeric CLEC4D/CLECSF8
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338339
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.