Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4E Antibody (16E3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP149311A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP149311A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP149311A32 Supplier Novus Biologicals Supplier No. NBP149311AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4E Monoclonal specifically detects CLEC4E in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CLEC4E
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 16000
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9
Gene Symbols CLEC4E
Host Species Mouse
Immunogen Recombinant human CLEC4E (41-219aa) purified from E. coli (NP_055173).
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26253
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.