Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4E Antibody, Novus Biologicals™
SDP

Catalog No. NBP227093SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227093SS 0.025 mg
NBP227093 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227093SS Supplier Novus Biologicals Supplier No. NBP227093SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLEC4E Polyclonal specifically detects CLEC4E in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen CLEC4E
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9ULY5
Gene Alias CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9
Gene Symbols CLEC4E
Host Species Rabbit
Immunogen This antibody was raised against CLEC4E
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26253
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, orangutan, and white-cheeked gibbon, 94% in crab-eating macaque, rhesus monkey, and sheep, 88% in cow, rat, and new world monkey, 82% in panda, dog, horse, and african bush elephant, 76% in guinea pig, and naked mole rat, 70% in mouse, and rabbit, 64% in Chinese hamster, opossum, and platypus.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.