Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4E Antibody (OTI2A8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271793A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271793A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271793A700 Supplier Novus Biologicals Supplier No. NBP271793AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4E Monoclonal specifically detects CLEC4E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CLEC4E
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2A8
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9
Gene Symbols CLEC4E
Host Species Mouse
Immunogen Full length recombinant protein of human CLEC4E (NP_055173) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26253.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.