Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4F/CLECSF13 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169537 25 μg
NB169536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB169537 Supplier Novus Biologicals Supplier No. NBP31723025UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLEC4F/CLECSF13 Polyclonal antibody specifically detects CLEC4F/CLECSF13 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen CLEC4F/CLECSF13
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13, C-type lectin 13, C-type lectin domain family 4, member F
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RVDNVNSQLQVLGDHLGNTNADIQMVKGVLKDATTLSLQTQMLRSSLEGTNAEIQRLKEDLEKADALTFQTLNFLKSSLENTSIELHVLSRGLENANSEIQMLNASLETANTQ
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165530
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.