Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (14N8D7), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP227087W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227087W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227087W Supplier Novus Biologicals Supplier No. NBP227087AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC9a Monoclonal specifically detects CLEC9a in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.

Specifications

Antigen CLEC9a
Applications Western Blot
Classification Monoclonal
Clone 14N8D7
Conjugate Alexa Fluor 405
Dilution Western Blot
Gene Accession No. Q6UXN8
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Gene Symbols CLEC9A
Host Species Mouse
Immunogen A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in Human, Orangutan, Chimp and Rhesus Monkey, 80% in Dog, pand and Horse, 73% in Bovine and 60% homologous in Mouse. CLEC9A is a glycoprotein visualized by western blot between ∽25-45 kD.
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.