Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (1F6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB384133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384133 Supplier Novus Biologicals Supplier No. NBP308840G
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CLEC9a
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 1F6
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Host Species Rat
Immunogen RBL-2H3 cells expressing Mouse CLEC9a fused to an HA epitope.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.