Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (2456A), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311491 Supplier Novus Biologicals Supplier No. FAB67762PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CLEC9a
Applications Flow Cytometry
Classification Monoclonal
Clone 2456A
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CLEC9a, Lys57-Ile264, Accession # EDK99924
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.