Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (30L2), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP221999Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221999Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221999Z Supplier Novus Biologicals Supplier No. NBP221999AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CLEC9a
Applications Flow Cytometry
Classification Monoclonal
Clone 30L2
Conjugate Alexa Fluor 700
Dilution Flow Cytometry
Gene Accession No. Q6UXN8
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Gene Symbols CLEC9A
Host Species Mouse
Immunogen Human recombinant CLEC9a
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.