Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9a Antibody (7H11) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB375762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375762 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375762 Supplier Novus Biologicals Supplier No. FAB67761Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen CLEC9a
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 7H11
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341
Host Species Rat
Immunogen RBL-2H3 rat basophilic leukemia cell line expressing mouse CLEC9a fused to an HA epitope, Accession # Q8BRU4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.