Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clenbuterol Antibody (1F8B10B7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002193 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002193 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002193 Supplier Novus Biologicals Supplier No. NB1002193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clenbuterol Monoclonal specifically detects Clenbuterol in Non-species specific samples. It is validated for ELISA, In vitro Assay, Surface Plasmon Resonance.

Specifications

Antigen Clenbuterol
Applications ELISA, In vitro Assay, Surface Plasmon Resonance
Classification Monoclonal
Clone 1F8B10B7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:10000, In vitro assay
Gene Alias 1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol, 37148-27-9, 4-Amino-3,5-dichloro-alpha-(((1,1-dimethylethyl)amino)methyl)benzenemethanol, Clenbuterolum, NAB 365, Planipart
Host Species Mouse
Immunogen Clenbuterol- KLH.
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Clenbuterol (1F8B10B7)
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.