Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC4 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB063743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063743 Supplier Novus Biologicals Supplier No. NBP299969AF532

Rabbit Polyclonal Antibody

CLIC4 Polyclonal specifically detects CLIC4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CLIC4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias chloride intracellular channel 4, chloride intracellular channel 4 like, chloride intracellular channel protein 4, CLIC4L, DKFZp566G223, FLJ38640, H1, huH1, Intracellular chloride ion channel protein p64H1, MTCLIC, p64H1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CLIC4 (Q9Y696-1; Ala 2-Lys 253).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25932
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.