Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC5 Antibody (CLIC5-02), HRP, Novus Biologicals™
SDP

Catalog No. NB002229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002229 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002229 Supplier Novus Biologicals Supplier No. NBP262197H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLIC5 Monoclonal specifically detects CLIC5 in Human samples. It is validated for Western Blot.

Specifications

Antigen CLIC5
Applications Western Blot
Classification Monoclonal
Clone CLIC5-02
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias chloride intracellular channel 5, chloride intracellular channel 5A, chloride intracellular channel protein 5, CLIC5B, dJ447E21.4, FLJ90663, MST130, MSTP130
Gene Symbols CLIC5
Host Species Mouse
Immunogen KLH-conjugated peptide corresponding to the amino acids 160-173 of human CLIC5a
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 53405
Test Specificity The antibody TSH-51 reacts with human thyroid stimulating hormone (hTSH, thyrotropin), a glycoprotein hormone produced by the anterior pituitary gland cells in response to signals from the hypothalamus gland in the brain. Following cross-reactivity expressed as binding of labelled hormone (% of total) was determined by solid phase RIA with excess of the antibody TSH-51: hTSH (68.6), hCG (0.03), hLH (2.99), hFSH (0.66).
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.