Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18008920UL
Description
CLIC6 Polyclonal specifically detects CLIC6 in Human samples. It is validated for Western Blot.Specifications
| CLIC6 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_444507 | |
| CLIC6 | |
| Synthetic peptide directed towards the middle region of human CLIC6. Peptide sequence RREDGEASEPRALGQEHDITLFVKAGYDGESIGNCPFSQRLFMILWLKGV. | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS & 2% Sucrose. with No Preservative | |
| chloride intracellular channel 6, parchorin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54102 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction