Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | CLIC6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18008920
![]() |
Novus Biologicals
NBP18008920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180089
![]() |
Novus Biologicals
NBP180089 |
100 μL |
Each for $487.50
|
|
|||||
Description
CLIC6 Polyclonal specifically detects CLIC6 in Human samples. It is validated for Western Blot.Specifications
| CLIC6 | |
| Polyclonal | |
| Rabbit | |
| NP_444507 | |
| 54102 | |
| Synthetic peptide directed towards the middle region of human CLIC6. Peptide sequence RREDGEASEPRALGQEHDITLFVKAGYDGESIGNCPFSQRLFMILWLKGV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| chloride intracellular channel 6, parchorin | |
| CLIC6 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title