Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLM-4 (CD300c) Rat anti-Mouse, Unconjugated, Clone: TX52, MilliporeSigma™
Rat Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | CLM-4 (CD300c) |
---|---|
Clone | TX52 |
Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23282MI
|
MilliporeSigma
MABF232825UG |
25 μg |
Each for $182.31
|
|
|||||
MABF23281MI
|
MilliporeSigma
MABF2328100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
CLM-4 (CD300c) Monoclonal antibody specifically detects CLM-4 (CD300c) in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting.Specifications
CLM-4 (CD300c) | |
Flow Cytometry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
CMRF-35-like molecule 4;CLM-4;CD300C molecule 2;Dendritic cell-derived Ig-like receptor 1;DIgR1;Immunoglobulin superfamily member 7;IgSF7;Leukocyte mono-Ig-like receptor 2;Myeloid-associated immunoglobulin-like receptor 2;MAIR-2;MAIR-II | |
Ba/F3 cells transfected with mouse CMRF-35-like molecule 4 (CD300c). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼25 and 33 kDa observed; 25.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
TX52 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_598919 | |
Cd300c2, Cd300c, Clm4, Igsf7, Lmir2 | |
IgG2b κ | |
Protein G Purified | |
Clone TX52 is a rat monoclonal antibody that specifically detects murine CMRF-35-like molecule 4 (CD300c). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title