Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLM-9 (CD300g) Rat anti-Mouse, Unconjugated, Clone: ZAQ2, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT1527100UL
Description
CLM-9 (CD300g) Monoclonal antibody specifically detects CLM-9 (CD300g) in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications
CLM-9 (CD300g) | |
Monoclonal | |
Unconjugated | |
CMRF35-like molecule 9;CD300 antigen-like family member G;Nepmucin;CD300g;CD300LG | |
Rat | |
∼32 kDa observed; 36.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μL | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
ZAQ2 | |
NP_082263 | |
Cd300lg, Clm9, D11Ertd736e | |
Extracellular domain of mouse CLM-9 (Nepmucin) protein fused with the Fc portion of human IgG. | |
Protein G Purified | |
Inflammation and Immunology | |
Clone ZAQ2 is a rat monoclonal antibody that detects murine CMRF35-like molecule 9 (CLM-9). | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction