Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLM-9 (CD300g) Rat anti-Mouse, Unconjugated, Clone: ZAQ2, MilliporeSigma™
Rat Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | CLM-9 (CD300g) |
---|---|
Clone | ZAQ2 |
Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15272MI
|
MilliporeSigma
MABT152725UL |
25 μL |
Each for $182.31
|
|
|||||
MABT15271MI
|
MilliporeSigma
MABT1527100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
CLM-9 (CD300g) Monoclonal antibody specifically detects CLM-9 (CD300g) in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.Specifications
CLM-9 (CD300g) | |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
CMRF35-like molecule 9;CD300 antigen-like family member G;Nepmucin;CD300g;CD300LG | |
Extracellular domain of mouse CLM-9 (Nepmucin) protein fused with the Fc portion of human IgG. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼32 kDa observed; 36.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
ZAQ2 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_082263 | |
Cd300lg, Clm9, D11Ertd736e | |
IgG2a κ | |
Protein G Purified | |
Clone ZAQ2 is a rat monoclonal antibody that detects murine CMRF35-like molecule 9 (CLM-9). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title