Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLOCK Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB059681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB059681 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB059681 Supplier Novus Biologicals Supplier No. NBP299290AF350

Rabbit Polyclonal Antibody

CLOCK Polyclonal specifically detects CLOCK in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CLOCK
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of mouse CLOCK.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 9575
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.