Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLOCK Rabbit anti-Human, Mouse, Rat, Clone: 3D8H6, Novus Biologicals™
SDP

Catalog No. NB168404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168404 20 μg
NB168403 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168404 Supplier Novus Biologicals Supplier No. NBP31665620UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CLOCK Monoclonal antibody specifically detects CLOCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen CLOCK
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 3D8H6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CLOCK (O15516).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 9575
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.