Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (+)-Clopidogrel hydrogen sulfate
GREENER_CHOICE

Catalog No. p-4783656
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective P2Y12 antagonist

Selective, high affinity P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity. Active enantiomer of (±)-clopidogrel hydrochloride (Cat. No. 2490).

Chemical Identifiers

CAS 120202-66-6
Molecular Formula C16H18ClNO6S2

Specifications

Quantity 10 mg
Formula Weight 419.9
Purity >99%
Molecular Weight (g/mol) 419.89
Chemical Name or Material (+)-Clopidogrel hydrogen sulfate
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.