Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Botulinum A Toxoid Antibody (KBA468), Novus Biologicals™
SDP

Catalog No. NB10073114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073114 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073114 Supplier Novus Biologicals Supplier No. NB10073114
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clostridium Botulinum A Toxoid Monoclonal specifically detects Clostridium Botulinum A Toxoid in Bacteria samples. It is validated for Sandwich ELISA.

Specifications

Antigen Clostridium Botulinum A Toxoid
Applications Sandwich ELISA
Classification Monoclonal
Clone KBA468
Conjugate Unconjugated
Dilution Sandwich ELISA 1:100-1:2000
Gene Alias Atx, Bna, BoNT/A, Bontoxilysin A, BotA, BOTOX, Botulinum neurotoxin type A, Botulism A toxoid
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with C. botulinum A toxoid.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity C. botulinum A toxoid. Cross-reactivity in indirect ELISA: B toxoid (1.0 %) and E toxoid (1.2 %).
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.