Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Botulinum Toxin A Antibody, Novus Biologicals™
SDP

Catalog No. NB11017098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11017098 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11017098 Supplier Novus Biologicals Supplier No. NB11017098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clostridium Botulinum Toxin A Polyclonal specifically detects Clostridium Botulinum Toxin A in Bacteria samples. It is validated for Western Blot, ELISA, Dot Blot.

Specifications

Antigen Clostridium Botulinum Toxin A
Applications Western Blot, ELISA, Dot Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:10000, Dot Blot 1:100-1:2000
Formulation 1X PBS (pH 7.2) containing 20% Glycerol with No Preservative
Gene Alias Atx, Bna, BoNT/A, Bontoxilysin A, BotA, BOTOX, Botulinum neurotoxin type A, Botulism A toxoid
Host Species Rabbit
Immunogen Type A Neurotoxin
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Clostridium botulinum Type A Neurotoxin.
Target Species Bacteria
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.