Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Difficile Toxin A Antibody, Novus Biologicals™
SDP

Catalog No. NB017481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017481 0.1 mL
NB10062473 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017481 Supplier Novus Biologicals Supplier No. NB100624730.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clostridium Difficile Toxin A Polyclonal specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Clostridium Difficile Toxin A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P16154
Gene Alias tcdA, toxA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino acids 2123 - 2132 (CTIDGKKYYFN) of the native molecule conjugated to Keyhole limpet Haemocyanin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62473 is specific for Clostridium difficile Toxin A, a gram-positive motile bacterium which is the leading cause of diarrhoea in developed countries. The incidence of disease in humans varies greatly with age, spore density, and the administration of antibiotics. Toxin A (TcdA) is a large exotoxin which is an essential virulence factor for C. difficile infection. TcdA has a molecular weight of 308kD and is a glucosyltransferase, which irreversibly inactivates the Ras family of small GTPases to disrupt many important signalling pathways.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.