Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Difficile Toxin B Antibody (5156), Novus Biologicals™
SDP

Catalog No. NBP121511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121511 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121511 Supplier Novus Biologicals Supplier No. NBP121511

Mouse Monoclonal Antibody

Clostridium Difficile Toxin B Monoclonal specifically detects Clostridium Difficile Toxin B in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Clostridium Difficile Toxin B
Applications ELISA
Classification Monoclonal
Clone 5156
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:20-1:200
Gene Alias tcdA; toxB
Host Species Mouse
Immunogen The immunogen is specific for Clostridium difficile toxin B.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NBP1-21511 is specific for Clostridium difficile toxin B. No reaction is observed with toxin A.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.