Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium perfringens Neuraminidase Recombinant Rabbit Monoclonal Antibody (112), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$293.50 - $565.00
Specifications
Antigen | Clostridium perfringens Neuraminidase |
---|---|
Clone | 112 |
Concentration | 1 mg/mL |
Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543473
|
Invitrogen™
MA543473 |
50 μL |
Each for $293.50
|
|
|||||
PIMA543055
|
Invitrogen™
MA543055 |
100 μL |
Each for $565.00
|
|
|||||
Description
Specificity: Clostridium Perfringens Neuraminidase
Sialidase, also known as Neuraminidase, is a secreted protein from bacterial species Clostridium perfringens that catalyses hydrolysis of glycosidic linkages on sialic acid residues in many biomolecules. It is suggested to be involved in the pathogenesis of microbial infections.Specifications
Clostridium perfringens Neuraminidase | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
Recombinant Clostridium Perfringens Neuraminidase protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
112 | |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Bacteria | |
Sialidase | |
IgG | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title