Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Tetani Toxoid Antibody (6E7), FITC, Novus Biologicals™
SDP

Catalog No. NB267587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB267587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB267587 Supplier Novus Biologicals Supplier No. NBP223554F

Mouse Monoclonal Antibody

Clostridium Tetani Toxoid Monoclonal antibody specifically detects Clostridium Tetani Toxoid in Bacteria samples. It is validated for ELISA

Specifications

Antigen Clostridium Tetani Toxoid
Applications ELISA
Classification Monoclonal
Clone 60000000
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX
Host Species Mouse
Immunogen Tetanus toxoid adsorbed to aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.