Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Tetani Toxoid Mouse anti-Bacteria, Clone: 10.D6, Novus Biologicals™
SDP

Catalog No. NBP22356201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22356201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22356201 Supplier Novus Biologicals Supplier No. NBP2235620.1ML

Mouse Monoclonal Antibody

Clostridium Tetani Toxoid Monoclonal specifically detects Clostridium Tetani Toxoid in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Clostridium Tetani Toxoid
Applications ELISA
Classification Monoclonal
Clone 10.D6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX
Gene Symbols TETX
Host Species Mouse
Immunogen Tetanus toxoid adsorbed to aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Clone 10.D6 binds to both tetanus toxin and tetanus toxoid
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.