Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Clozapine

Click to view available options
Quantity:
50 mg
500 mg
Description
Clozapine is an atypical antipsychotic drug, with a much lower tendency to cause extrapyramidal side effects than conventional neuroleptics. Displays a broad range of pharmacological actions; the antipsychotic effects are thought to be mediated principally by 5-HT2A/2C and dopamine receptor blockade (Ki values are 21, 170, 170, 230 and 330 nM for D4, D3, D1, D2 and D5 receptors respectively).
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 326.83 |
Purity | >99% |
Molecular Weight (g/mol) | 326.828 |
Chemical Name or Material | Clozapine |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction