Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLP24/TMEM204 Antibody (652424), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311515 Supplier Novus Biologicals Supplier No. FAB6747PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLP24/TMEM204 Monoclonal antibody specifically detects CLP24/TMEM204 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CLP24/TMEM204
Applications Flow Cytometry
Classification Monoclonal
Clone 652424
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias C16orf30chromosome 16 open reading frame 30, Claudin-like protein 24, CLP24MGC111564, FLJ20898, transmembrane protein 204
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CLP24, Accession # Q9BSN7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79652
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.