Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPP Antibody (OTI1F6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB271363APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271363APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271363APC Supplier Novus Biologicals Supplier No. NBP271363APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLPP Monoclonal specifically detects CLPP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen CLPP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1F6
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP-dependent protease ClpAP, proteolytic subunit, human, ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli)homolog, ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E.coli), ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog, ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E.coli), EC 3.4.21.92, Endopeptidase Clp, putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Symbols CLPP
Host Species Mouse
Immunogen Full length recombinant protein of human CLPP (NP_006003) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8192.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.