Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPP Rabbit anti-Human, Mouse, Rat, Clone: S02-1E5, Novus Biologicals™
SDP

Catalog No. NB188381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188381 25 μg
NB188380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188381 Supplier Novus Biologicals Supplier No. NBP31501425UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CLPP Monoclonal antibody specifically detects CLPP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen CLPP
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S02-1E5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 10 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias ATP-dependent protease ClpAP, proteolytic subunit, human, ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli)homolog, ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E.coli), ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog, ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E.coli), EC 3.4.21.92, Endopeptidase Clp, putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Host Species Rabbit
Immunogen A synthetic peptide of human CLPP (Uniprot#: Q16740)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8192
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.